Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081424.1 | NP_001074893.1 | 3 | 693 | ||
GenBank mRNA | AK033068.1 | - | 3 | 693 | ||
BC147245.1 | - | 3 | 408 | 9056293 | ||
BC147246.1 | - | 3 | 408 | 9056310 | ||
BC151033.1 | - | 3 | 408 | 9087946 | ||
BC151035.1 | - | 3 | 408 | 9087948 |