Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015766.2 | NP_056581.1 | 5 | 897 | ||
GenBank mRNA | AF013114.1 | - | 5 | 880 | ||
AK155023.1 | - | 5 | 860 | |||
AK155903.1 | - | 5 | 752 | |||
AW495532.1 | - | 1 | 287 | |||
BC008209.1 | - | 5 | 845 | 3154957 | ||
BI134394.1 | - | 1 | 288 |