Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037539.2 | NP_001032628.1 | 3 | 379 | ||
NM_001170419.1 | NP_001163890.1 | 2 | 253 | |||
NM_015768.2 | NP_056583.1 | 4 | 442 | |||
GenBank mRNA | AF182064.1 | - | 4 | 442 | ||
AF182066.1 | - | 3 | 379 | |||
AF487280.1 | - | 3 | 370 | |||
AK015462.1 | - | 4 | 404 | |||
BB614931.1 | - | 2 | 245 | |||
BC116911.1 | - | 4 | 411 | 40126230 | ||
BC144863.1 | - | 3 | 360 | 9053411 |