Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015798.3 | NP_056613.1 | 10 | 1513 | ||
GenBank mRNA | AF176530.1 | - | 10 | 1497 | ||
AK082809.1 | - | 10 | 1534 | |||
AK163224.1 | - | 10 | 1514 | |||
AK166591.1 | - | 10 | 1515 | |||
AK166599.1 | - | 10 | 1516 | |||
AK166918.1 | - | 10 | 2379 | |||
AK167118.1 | - | 10 | 1515 | |||
AK167270.1 | - | 10 | 1513 |