Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282061.1 | NP_001268990.1 | 16 | 1384 | ||
NM_015812.3 | NP_056627.1 | 16 | 1666 | |||
GenBank mRNA | AF061933.1 | - | 13 | 1111 | ||
AK044067.1 | - | 2 | 55 | |||
AK052527.1 | - | 16 | 1667 | |||
AK053087.1 | - | 16 | 1385 | |||
BC038616.1 | - | 14 | 1211 | |||
BC125029.1 | - | 16 | 1370 | 40062355 | ||
BC125030.1 | - | 16 | 1370 | 40062356 |