Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016712.3 | NP_057921.1 | 5, 6 | 601 | ||
GenBank mRNA | AF177174.1 | - | 4, 5 | 520 | ||
AK144408.1 | - | 5, 6 | 602 | |||
AK144535.1 | - | 5, 6 | 602 | |||
BC068020.1 | - | 4, 5 | 537 | 30291625 |