Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nckap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290745.1 | NP_001277674.1 | 19, 20 | 2106 | ||
NM_016965.3 | NP_058661.1 | 18, 19 | 2088 | |||
GenBank mRNA | AJ534525.1 | - | 18, 19 | 1871 | ||
AK122318.1 | - | 17, 18 | 1909 | |||
AK209671.1 | - | 3, 4 | 275 | |||
BC060197.1 | - | 18, 19 | 2066 | |||
BC127067.1 | - | 19, 20 | 1888 | |||
X61453.1 | - | 10, 11 | 1010 |