Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nckap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290745.1 | NP_001277674.1 | 29, 30 | 3306 | ||
NM_016965.3 | NP_058661.1 | 28, 29 | 3288 | |||
GenBank mRNA | AJ534525.1 | - | 28, 29 | 3071 | ||
AK122318.1 | - | 27, 28 | 3109 | |||
AK143049.1 | - | 5, 6 | 446 | |||
BC003962.1 | - | 8, 9 | 810 | |||
BC032206.1 | - | 7, 8 | 650 | |||
BC060197.1 | - | 28, 29 | 3266 | |||
BC127067.1 | - | 29, 30 | 3088 |