Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190449.1 | NP_001177378.1 | 2, 3 | 599 | ||
NM_016765.3 | NP_058045.1 | 2, 3 | 599 | |||
GenBank mRNA | AF004106.1 | - | 2, 3 | 533 | ||
BC003328.1 | - | 2, 3 | 549 | 3158175 |