Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rundc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252347.1 | NP_001239276.1 | 7 | 1044 | ||
NM_016759.4 | NP_058039.1 | 7 | 1044 | |||
GenBank mRNA | AK002856.1 | - | 3 | 342 | ||
BC022745.1 | - | 5 | 769 | |||
BC046319.1 | - | 7 | 944 | 6391042 | ||
U73941.1 | - | 7 | 771 |