Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024204.6 | NP_077166.4 | 6 | 788 | ||
GenBank mRNA | AK012869.1 | - | 6 | 801 | ||
AK017360.1 | - | 6 | 789 | |||
AK017408.1 | - | 6 | 673 | |||
AK037242.1 | - | 6 | 671 | |||
BC003460.1 | - | 6 | 649 | 2648395 |