Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033128.3 | NP_001028300.1 | 16, 17 | 1717 | ||
GenBank mRNA | AK037753.1 | - | 16, 17 | 1703 | ||
AK045850.1 | - | 16, 17 | 1715 | |||
AK163815.1 | - | 7, 8 | 885 | |||
BC076577.1 | - | 16, 17 | 1706 |