Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgsnat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029884.1 | NP_084160.1 | 15, 16 | 1638 | ||
GenBank mRNA | AK035264.1 | - | 15, 16 | 1571 | ||
AK149883.1 | - | 15, 16 | 1642 | |||
AK152926.1 | - | 15, 16 | 1603 | |||
AK159649.1 | - | 15, 16 | 1567 | |||
AK160068.1 | - | 15, 16 | 1561 | |||
AK209731.1 | - | 2, 3 | 80 | |||
BC024084.1 | - | 15, 16 | 1545 | 5101064 |