Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgsnat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029884.1 | NP_084160.1 | 3 | 429 | ||
GenBank mRNA | AK035264.1 | - | 3 | 362 | ||
AK149883.1 | - | 3 | 433 | |||
AK152926.1 | - | 3 | 394 | |||
AK159649.1 | - | 3 | 358 | |||
AK160068.1 | - | 3 | 352 |