Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgsnat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029884.1 | NP_084160.1 | 4 | 561 | ||
GenBank mRNA | AK035264.1 | - | 4 | 494 | ||
AK152926.1 | - | 4 | 526 | |||
AK159649.1 | - | 4 | 490 | |||
AK160068.1 | - | 4 | 484 | |||
AK191575.1 | - | 259 | ||||
BC024084.1 | - | 4 | 468 | 5101064 |