Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033525.3 | NP_001028697.2 | 3 | 918 | ||
GenBank mRNA | AK036882.1 | - | 3 | 873 | ||
AK132480.1 | - | 3 | 919 | |||
AK163847.1 | - | 3 | 767 | |||
AK171038.1 | - | 3 | 912 | |||
BC115791.1 | - | 3 | 624 | 40100611 | ||
BC115915.1 | - | 3 | 618 | 40100605 | ||
DQ185135.1 | - | 3 | 879 |