Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026187.5 | NP_080463.3 | 6 | 1167 | ||
GenBank mRNA | AK004948.2 | - | 4 | 1597 | ||
AK156255.1 | - | 6 | 1131 | |||
AK157405.1 | - | 6 | 1131 | |||
BC039910.1 | - | 5 | 1473 | |||
BC071240.1 | - | 6 | 1097 | |||
BC150879.1 | - | 6 | 1117 | 9087790 | ||
BU054541.1 | - | 5 | 568 |