Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D1Ertd622e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133825.3 | NP_598586.1 | 3 | 640 | ||
GenBank mRNA | AK135656.1 | - | 3 | 586 | ||
AK137265.1 | - | 3 | 1399 | |||
AK152370.1 | - | 3 | 612 | |||
AK162223.1 | - | 3 | 584 | |||
AK168549.1 | - | 3 | 615 | |||
BC019375.1 | - | 2 | 497 | 5008599 | ||
BC023951.1 | - | 3 | 609 | 5320263 | ||
BC034088.1 | - | 3 | 1103 |