Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198831.2 | NP_942128.2 | 6 | 522 | ||
GenBank mRNA | AK007653.1 | - | 6 | 792 | ||
AK171917.1 | - | 6 | 495 | |||
BC037190.1 | - | 6 | 477 | 4168034 | ||
BC096562.1 | - | 4 | 319 | |||
BC116358.1 | - | 5 | 392 | 40099962 | ||
BC116359.1 | - | 5 | 389 | 40099963 | ||
BC128487.1 | - | 5 | 392 | 40099964 | ||
BC128488.1 | - | 5 | 392 | 40099965 | ||
BG077486.2 | - | 5 | 356 | |||
BU604260.1 | - | 6 | 515 | |||
CD363582.1 | - | 6 | 453 | |||
DN174444.1 | - | 6 | 522 |