Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199020.1 | NP_001185949.1 | 9 | 1411 | ||
NM_021421.4 | NP_067396.3 | 8 | 1546 | |||
GenBank mRNA | AB041602.1 | - | 6 | 1079 | ||
AK077039.1 | - | 8 | 1493 | |||
AK088834.1 | - | 7 | 1179 | |||
AK135709.1 | - | 9 | 1408 | |||
AK158731.1 | - | 8 | 1562 | |||
AK166619.1 | - | 9 | 1410 | |||
AK169653.1 | - | 7 | 1130 | |||
AK199712.1 | - | 2 | 57 | |||
AK214324.1 | - | 2 | 57 | |||
AK216183.1 | - | 2 | 57 | |||
BC024907.1 | - | 8 | 1506 | 2812124 |