Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029990.1 | NP_001025161.1 | 7, 8 | 801 | ||
GenBank mRNA | AK009576.1 | - | 7, 8 | 714 | ||
AK087101.1 | - | 7, 8 | 733 | |||
BC138944.1 | - | 7, 8 | 754 | 8861966 |