Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026383.3 | NP_080659.1 | 3 | 780 | ||
GenBank mRNA | AK011192.1 | - | 3 | 758 | ||
AK012073.1 | - | 3 | 764 | |||
AK013163.1 | - | 3 | 764 | |||
AK018384.1 | - | 3 | 728 | |||
AK031048.1 | - | 1778 | ||||
AK075816.1 | - | 3 | 764 | |||
AK077403.1 | - | 3 | 645 | |||
AK207137.1 | - | 1 | 376 | |||
AK207206.1 | - | 1 | 47 | |||
AK210203.1 | - | 1 | 376 | |||
AK215332.1 | - | 1 | 30 | |||
BC006598.1 | - | 3 | 753 | 3965195 |