Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177778.2 | NP_001171249.1 | 13 | 2160 | ||
NM_001177779.1 | NP_001171250.1 | 6 | 1061 | |||
NM_001177780.1 | NP_001171251.1 | 4 | 453 | |||
NM_001290402.1 | NP_001277331.1 | 12 | 2106 | |||
NM_016747.4 | NP_058027.1 | 13 | 2160 | |||
GenBank mRNA | AK122474.1 | - | 13 | 2159 | ||
AK145815.1 | - | 6 | 816 | |||
AK148464.1 | - | 4 | 432 | |||
AK159655.1 | - | 4 | 441 | |||
AK167365.1 | - | 4 | 440 | |||
BC060713.1 | - | 11 | 2138 | |||
BC094368.1 | - | 12 | 2101 | 6842105 | ||
CN698511.1 | - | 4 | 453 | |||
D87117.1 | - | 13 | 2134 |