Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgrmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016783.3 | NP_058063.2 | 2, 3 | 535 | ||
GenBank mRNA | AF042491.1 | - | 2, 3 | 486 | ||
AK153272.1 | - | 2, 3 | 552 | |||
AK159101.1 | - | 2, 3 | 552 | |||
AK159345.1 | - | 2, 3 | 552 | |||
AK159599.1 | - | 2, 3 | 552 | |||
AK162315.1 | - | 2, 3 | 555 | |||
AK166240.1 | - | 2, 3 | 552 | |||
AK167655.1 | - | 2, 3 | 552 | |||
AK168387.1 | - | 2, 3 | 551 | |||
AK168882.1 | - | 2, 3 | 552 | |||
AK169063.1 | - | 3, 4 | 495 | |||
AK193145.1 | - | 1, 2 | 123 | |||
AK208626.1 | - | 1, 2 | 123 | |||
AK214321.1 | - | 1 | 123 | |||
BC006016.1 | - | 2, 3 | 535 | 3591536 |