Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdcbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098227.1 | NP_001091697.1 | 2, 3 | 144 | ||
NM_016807.2 | NP_058087.2 | 2, 3 | 144 | |||
GenBank mRNA | AF003693.1 | - | 2, 3 | 136 | ||
AF077527.1 | - | 2, 3 | 103 | |||
AK014678.1 | - | 118 | ||||
AK028429.1 | - | 2, 3 | 117 | |||
AK033412.1 | - | 2, 3 | 142 | |||
AK146085.1 | - | 2, 3 | 116 | |||
AK150278.1 | - | 2, 3 | 109 | |||
AK150321.1 | - | 2, 3 | 112 | |||
AK150544.1 | - | 2, 3 | 116 | |||
AK150623.1 | - | 2, 3 | 120 | |||
AK150661.1 | - | 2, 3 | 112 | |||
AK150707.1 | - | 2, 3 | 115 | |||
AK150721.1 | - | 2, 3 | 124 | |||
AK150736.1 | - | 2, 3 | 112 | |||
AK150801.1 | - | 2, 3 | 114 | |||
AK150996.1 | - | 2, 3 | 116 | |||
AK151050.1 | - | 2, 3 | 116 | |||
AK152001.1 | - | 2, 3 | 115 | |||
AK152017.1 | - | 2, 3 | 116 | |||
AK152080.1 | - | 2, 3 | 123 | |||
AK152109.1 | - | 2, 3 | 116 | |||
AK152434.1 | - | 2, 3 | 116 | |||
AK152552.1 | - | 2, 3 | 120 | |||
AK152577.1 | - | 2, 3 | 115 | |||
AK152855.1 | - | 2, 3 | 116 | |||
AK153042.1 | - | 2, 3 | 116 | |||
AK153112.1 | - | 2, 3 | 120 | |||
AK159124.1 | - | 2, 3 | 112 | |||
AK159323.1 | - | 2, 3 | 90 | |||
AK159538.1 | - | 2, 3 | 117 | |||
AK159677.1 | - | 2, 3 | 123 | |||
AK165657.1 | - | 2, 3 | 114 | |||
AK167513.1 | - | 2, 3 | 112 | |||
AK168004.1 | - | 2, 3 | 119 | |||
AK168777.1 | - | 2, 3 | 112 | |||
AK169219.1 | - | 2, 3 | 121 | |||
AK169415.1 | - | 2, 3 | 150 | |||
AK169687.1 | - | 2, 3 | 116 | |||
AK171162.1 | - | 2, 3 | 116 | |||
AK191271.1 | - | 2, 3 | 84 | |||
AK215623.1 | - | 2, 3 | 84 | |||
BC019400.1 | - | 2, 3 | 107 | 4017442 | ||
BY779386.1 | - | 2, 3 | 145 |