Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016792.4 | NP_058072.2 | 3 | 444 | ||
GenBank mRNA | AF052660.1 | - | 3 | 273 | ||
AK029807.1 | - | 3 | 445 | |||
AK051698.1 | - | 3 | 419 | |||
AK144211.1 | - | 3 | 401 | |||
AK150326.1 | - | 3 | 407 | |||
AK150887.1 | - | 3 | 413 | |||
AK150983.1 | - | 3 | 413 | |||
AK152101.1 | - | 3 | 413 | |||
AK167954.1 | - | 3 | 415 | |||
AK168712.1 | - | 3 | 408 | |||
AK212711.1 | - | 1 | 26 | |||
BC061123.1 | - | 3 | 471 | 6596299 |