Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec61a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016906.4 | NP_058602.1 | 6, 7 | 578 | ||
GenBank mRNA | AB032902.1 | - | 6, 7 | 590 | ||
AF145253.1 | - | 6, 7 | 500 | |||
AF222743.1 | - | 6, 7 | 500 | |||
AK054318.1 | - | 1, 2 | 1414 | |||
AK088473.1 | - | 6, 7 | 579 | |||
AK153055.1 | - | 6, 7 | 568 | |||
AK159349.1 | - | 6, 7 | 566 | |||
AK167484.1 | - | 6, 7 | 508 | |||
AK197110.1 | - | 2, 3 | 190 | |||
BC003707.1 | - | 6, 7 | 504 | 3592772 | ||
BC006020.1 | - | 2, 3 | 163 |