Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nono | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252518.1 | NP_001239447.1 | 7 | 997 | ||
NM_023144.2 | NP_075633.2 | 7 | 1000 | |||
GenBank mRNA | AK004830.1 | - | 7 | 963 | ||
AK028338.1 | - | 7 | 965 | |||
AK033201.1 | - | 7 | 926 | |||
AK145179.1 | - | 7 | 973 | |||
AK161152.1 | - | 7 | 984 | |||
AK165855.1 | - | 7 | 959 | |||
AK167297.1 | - | 7 | 931 | |||
AK169202.1 | - | 7 | 963 | |||
AK169303.1 | - | 7 | 984 | |||
AK185402.1 | - | 3 | 340 | |||
AK189617.1 | - | 3 | 340 | |||
AK213541.1 | - | 3 | 340 | |||
BC005465.1 | - | 7 | 959 | 3600935 | ||
BC083074.1 | - | 5 | 837 | 6390386 | ||
CT010241.1 | - | 5 | 819 | |||
S64860.1 | - | 7 | 947 |