Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nono | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252518.1 | NP_001239447.1 | 7 | 1038 | ||
NM_023144.2 | NP_075633.2 | 7 | 1041 | |||
GenBank mRNA | AK004830.1 | - | 7 | 1004 | ||
AK028338.1 | - | 7 | 1006 | |||
AK145179.1 | - | 7 | 1014 | |||
AK161152.1 | - | 7 | 1025 | |||
AK165855.1 | - | 7 | 1000 | |||
AK167297.1 | - | 7 | 972 | |||
AK169202.1 | - | 7 | 1004 | |||
AK169303.1 | - | 7 | 1025 | |||
AK183390.1 | - | 3 | 224 | |||
AK185402.1 | - | 3 | 381 | |||
AK189617.1 | - | 3 | 381 | |||
BC005465.1 | - | 7 | 1000 | 3600935 | ||
BC083074.1 | - | 5 | 878 | 6390386 | ||
CT010241.1 | - | 5 | 860 | |||
S64860.1 | - | 7 | 988 |