Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004177.2 | NP_001004177.2 | 1 | 2237 | ||
NM_017392.3 | NP_059088.2 | 1 | 2237 | |||
GenBank mRNA | AB028499.1 | - | 1 | 2240 |
Set Membership: |
G-protein coupled receptor |