Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286653.1 | NP_001273582.1 | 3 | 457 | ||
NM_001286654.1 | NP_001273583.1 | 3 | 334 | |||
NM_030598.2 | NP_085101.1 | 2 | 464 | |||
NM_207649.1 | NP_997532.1 | 3 | 590 | |||
GenBank mRNA | AB061524.1 | - | 2 | 402 | ||
AB061525.1 | - | 3 | 457 | |||
AF237791.1 | - | 2 | 204 | |||
AF237887.1 | - | 2 | 250 | |||
AK049006.1 | - | 2 | 403 | |||
AK136805.1 | - | 3 | 398 | |||
AK138768.1 | - | 3 | 502 | |||
AK160865.1 | - | 2 | 352 | |||
BB635249.1 | - | 3 | 502 | |||
BC047153.1 | - | 2 | 262 | 6491318 | ||
BC049096.1 | - | 2 | 464 | 6416901 | ||
BC062141.1 | - | 3 | 334 | 30290791 | ||
BF534085.1 | - | 437 | ||||
BY740155.1 | - | 3 | 597 |