Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252600.1 | NP_001239529.1 | 4 | 791 | ||
NM_001252601.1 | NP_001239530.1 | 4 | 791 | |||
NM_016850.3 | NP_058546.1 | 4 | 791 | |||
GenBank mRNA | AK002830.1 | - | 4 | 790 | ||
AK004349.1 | - | 4 | 791 | |||
AK079685.1 | - | 4 | 791 | |||
AK089869.1 | - | 4 | 791 | |||
AK150403.1 | - | 4 | 791 | |||
AK150608.1 | - | 4 | 791 | |||
AK151470.1 | - | 4 | 791 | |||
AK152401.1 | - | 4 | 791 | |||
AK152403.1 | - | 4 | 791 | |||
AK153054.1 | - | 4 | 791 | |||
AK155950.1 | - | 1 | 1304 | |||
AK158378.1 | - | 4 | 790 | |||
AK158403.1 | - | 4 | 790 | |||
AK159142.1 | - | 4 | 790 | |||
AK159883.1 | - | 4 | 791 | |||
BC138799.1 | - | 3 | 731 | 8861821 | ||
BC145422.1 | - | 3 | 731 | |||
DT923384.1 | - | 2 | 268 | |||
U73037.1 | - | 3 | 768 |