Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017401.2 | NP_059097.2 | 2 | 567 | ||
GenBank mRNA | AF176098.1 | - | 2 | 340 | ||
AJ251804.2 | - | 2 | 311 | |||
AK137747.1 | - | 1 | 1517 | |||
AK208265.1 | - | 2 | 142 | |||
BC056213.1 | - | 2 | 359 | 30024844 | ||
BC056996.1 | - | 1 | 1705 |