Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113517.1 | NP_001106989.1 | 8 | 1015 | ||
NM_001113518.1 | NP_001106990.1 | 7 | 804 | |||
NM_017402.4 | NP_059098.2 | 7 | 804 | |||
GenBank mRNA | AF247654.1 | - | 7 | 700 | ||
AF247655.1 | - | 5 | 421 | |||
AF343877.1 | - | 7 | 684 | |||
AK052545.1 | - | 7 | 775 | |||
AK129064.1 | - | 8 | 951 | |||
AK139156.1 | - | 8 | 944 | |||
AY220301.1 | - | 7 | 769 | |||
BC044838.1 | - | 7 | 789 | 3499604 | ||
U96634.1 | - | 5 | 494 |