Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016861.4 | NP_058557.2 | 6 | 908 | ||
GenBank mRNA | AF053367.2 | - | 6 | 824 | ||
AK076285.1 | - | 807 | ||||
AK088089.1 | - | 6 | 807 | |||
AK146225.1 | - | 6 | 818 | |||
AK146399.1 | - | 6 | 820 | |||
AK158173.1 | - | 6 | 807 |