Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acrbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127340.1 | NP_001120812.1 | 5 | 652 | ||
NM_016845.2 | NP_058541.2 | 5 | 652 | |||
GenBank mRNA | AK076691.1 | - | 5 | 617 | ||
AK132705.1 | - | 5 | 620 | |||
AV260707.2 | - | 1 | 40 | |||
BC011079.1 | - | 5 | 890 | 3984550 | ||
BM938496.1 | - | 4 | 425 | |||
D17573.1 | - | 5 | 604 | |||
D17574.1 | - | 5 | 656 |