Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017470.2 | NP_059498.2 | 3 | 339 | ||
GenBank mRNA | AB010031.1 | - | 3 | 317 | ||
AK002657.1 | - | 3 | 290 | |||
AK029618.1 | - | 2 | 150 | |||
AK132061.1 | - | 3 | 329 | |||
BC005426.1 | - | 3 | 268 | 3593932 | ||
BY012683.1 | - | 3 | 340 |