Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017470.2 | NP_059498.2 | 4 | 442 | ||
GenBank mRNA | AB010031.1 | - | 4 | 420 | ||
AK002657.1 | - | 4 | 393 | |||
AK029618.1 | - | 3 | 253 | |||
AK132061.1 | - | 4 | 432 | |||
BC005426.1 | - | 4 | 371 | 3593932 |