Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019397.3 | NP_062270.1 | 6, 7 | 851 | ||
GenBank mRNA | AJ245672.1 | - | 6, 7 | 853 | ||
AK053738.1 | - | 6, 7 | 848 | |||
AK078534.1 | - | 6, 7 | 829 | |||
BC117702.2 | - | 6, 7 | 837 | 40062530 |