Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnase2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019398.2 | NP_062271.2 | 1 | 506 | ||
GenBank mRNA | BC117776.2 | - | 1 | 506 | 40091395 | |
BC119505.1 | - | 1 | 299 | |||
BC119506.1 | - | 1 | 302 | 40053222 |