Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rragc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017475.2 | NP_059503.2 | 5 | 962 | ||
GenBank mRNA | AB017616.1 | - | 5 | 885 | ||
AK150792.1 | - | 5 | 917 | |||
AK159355.1 | - | 5 | 895 | |||
AK159434.1 | - | 5 | 917 | |||
AK166658.1 | - | 5 | 908 | |||
BC005417.1 | - | 5 | 910 | 3495022 | ||
BC037732.1 | - | 2 | 259 | |||
BC071245.1 | - | 5 | 1310 |