Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019403.3 | NP_062276.1 | 1, 2 | 227 | ||
GenBank mRNA | AK010792.1 | - | 1, 2 | 257 | ||
AK012729.1 | - | 1, 2 | 219 | |||
AK019289.1 | - | 1, 2 | 238 | |||
AK020514.2 | - | 1, 2 | 228 | |||
AK020515.2 | - | 1, 2 | 246 | |||
BC012031.1 | - | 1, 2 | 178 | |||
BC016449.1 | - | 1, 2 | 227 | 4191746 |