Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017461.2 | NP_059489.2 | 9 | 822 | ||
GenBank mRNA | BC140998.1 | - | 9 | 823 | 9055610 | |
BC145081.1 | - | 9 | 823 | 9053629 | ||
BX523183.1 | - | 2 | 138 | |||
M37030.1 | - | 9 | 826 |