Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnpnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019425.2 | NP_062298.1 | 5, 6 | 472 | ||
GenBank mRNA | AJ001006.1 | - | 5, 6 | 472 | ||
AK002466.1 | - | 5, 6 | 493 | |||
AK007647.1 | - | 5, 6 | 477 | |||
AK007722.1 | - | 5, 6 | 477 | |||
AK007764.1 | - | 5, 6 | 500 | |||
AK008566.1 | - | 5, 6 | 482 | |||
AK011098.1 | - | 5, 6 | 500 | |||
AK018499.1 | - | 5, 6 | 466 | |||
AK028693.1 | - | 5, 6 | 498 | |||
AK050266.1 | - | 5, 6 | 470 | |||
AK167870.1 | - | 5, 6 | 482 | |||
AK168074.1 | - | 5, 6 | 482 | |||
BC031116.1 | - | 5, 6 | 462 | 5065641 | ||
BC092047.1 | - | 5, 6 | 453 |