Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018750.3 | NP_061220.2 | 1, 2 | 484 | ||
GenBank mRNA | AF053959.1 | - | 1, 2 | 466 | ||
AK005059.2 | - | 1, 2 | 480 | |||
AK029321.1 | - | 1, 2 | 486 | |||
AK155534.1 | - | 1, 2 | 480 | |||
BC089605.1 | - | 1, 2 | 486 | 30121804 |