Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018817.2 | NP_061287.2 | 13 | 2298 | ||
GenBank mRNA | AF088884.1 | - | 13 | 2278 | ||
AK077878.1 | - | 13 | 2276 | |||
AK083488.1 | - | 13 | 2276 | |||
AK146767.1 | - | 13 | 2294 | |||
AK154020.1 | - | 13 | 2281 | |||
AK161570.1 | - | 12 | 3590 | |||
BC029078.1 | - | 13 | 2233 | 3992325 |