Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018755.2 | NP_061225.2 | 7 | 1296 | ||
NM_176073.4 | NP_788262.1 | 6 | 1206 | |||
GenBank mRNA | AF009513.1 | - | 6 | 1051 | ||
AF107832.1 | - | 3 | 175 | |||
AF107835.1 | - | 6 | 1174 | |||
AK032972.1 | - | 6 | 1071 | |||
AK075686.1 | - | 6 | 1137 | |||
AK135096.1 | - | 6 | 1193 | |||
AK137164.1 | - | 6 | 1080 | |||
AK152139.1 | - | 7 | 1174 | |||
AK152145.1 | - | 7 | 1174 | |||
BC010977.1 | - | 6 | 1191 | |||
BC037067.1 | - | 6 | 1053 | 4989536 |