Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018753.6 | NP_061223.2 | 3 | 556 | ||
GenBank mRNA | AF058797.1 | - | 3 | 555 | ||
AK002632.1 | - | 3 | 575 | |||
AK004872.1 | - | 3 | 557 | |||
AK011389.1 | - | 3 | 556 | |||
AK083367.1 | - | 3 | 546 | |||
AK144061.1 | - | 3 | 619 | |||
AK158932.1 | - | 3 | 459 | |||
AK178774.1 | - | 2 | 116 | |||
AK184712.1 | - | 2 | 116 | |||
AK186637.1 | - | 2 | 117 | |||
AK206680.1 | - | 1 | 87 | |||
AK209277.1 | - | 2 | 116 | |||
AK217062.1 | - | 1 | 99 | |||
AK219160.1 | - | 1 | 88 | |||
BC049070.1 | - | 390 | ||||
BC132121.1 | - | 3 | 410 | 40130398 | ||
BC138512.1 | - | 3 | 410 | 8861532 |