Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136260.1 | NP_001129732.1 | 22 | 2844 | ||
NM_001197147.1 | NP_001184076.1 | 22 | 2817 | |||
NM_018760.2 | NP_061230.2 | 22 | 2844 | |||
GenBank mRNA | AW124701.1 | - | 2 | 285 | ||
BC026592.1 | - | 13 | 1637 | |||
BC129863.1 | - | 22 | 2806 | |||
BC129864.1 | - | 22 | 2805 | |||
BC148292.2 | - | 22 | 2780 | 40110074 | ||
CB524421.1 | - | 7 | 705 | |||
HQ018820.1 | - | 19 | 2681 | |||
HQ204220.1 | - | 22 | 2788 | |||
HQ285250.1 | - | 19 | 2657 |