Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136260.1 | NP_001129732.1 | 7 | 844 | ||
NM_001197147.1 | NP_001184076.1 | 7 | 817 | |||
NM_018760.2 | NP_061230.2 | 7 | 844 | |||
GenBank mRNA | AF020195.1 | - | 7 | 807 | ||
AF141934.3 | - | 4 | 646 | |||
AK140443.1 | - | 7 | 1287 | |||
BC129863.1 | - | 7 | 806 | |||
BC129864.1 | - | 7 | 805 | |||
BC148292.2 | - | 7 | 780 | 40110074 | ||
HQ018820.1 | - | 4 | 681 | |||
HQ204220.1 | - | 7 | 788 | |||
HQ285250.1 | - | 4 | 657 |